Learn More
Interscience™ Set of 5 Teflon™ Nozzles
Product Code. 15530703
Change view
Click to view available options
Quantity:
5 Nozzles/Set
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15530703 5 Nozzles/Set Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15530703 Supplier Interscience™ Supplier No. 513024
  • ONLINE EXCLUSIVE PRICE
    2150.00 NOK
    2092.38 NOK / Pack of 1
    Save 57.62 NOK
    2% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Set of 5 nozzles and caps for dilutor tubings.
  • For tubings with 4.8 to 8 mm inner diameter
Specifications
Product Type Tubing Nozzle
Autoclavable Autoclavable
For Use With (Application) Applying Nozzles
For Use With (Equipment) DiluFlow, DiluFlow Pro, DiluFlow Elite, FlexiPump, FlexiPump Pro
Quantity 5 Nozzles/Set