Learn More
Labconco™ Service Fixture Kits
Product Code. 13141154 Shop All Labconco Corporation Products
Change view
Click to view available options
Product Type:
Cold Water (CW) GN Fixture Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
13141154 Cold Water (CW) GN Fixture Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13141154 Supplier Labconco™ Supplier No. 9827900
  • ONLINE EXCLUSIVE PRICE
    8560.00 NOK
    6908.49 NOK / Pack of 1
    Save 1651.51 NOK
    19% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Kit includes copper tubing, brass valve, knob handle and outlet.
  • Max. working pressure of 40 psi.
  • Kits mount on the right or left side of any Protector Premier, XStream, XL, or PVC Hood.
  • Each kit includes one remotely-controlled service fixture with valve and tubing, color-coded fixture knob.
Specifications
For Use With (Equipment) Protector XLE Laboratory Hood
Weight (English) 10 lb.
Weight (Metric) 4.5 Kg
Includes 0.37 in. dia. Brass Valve, Copper Tubing, Green Fixture Knob, Hose Connector
Product Type Cold Water (CW) GN Fixture Kit
For Use With (Application) Cold water service