Learn More
Greiner Bio-One VACUETTE™ Z Serum Blood Collection Tubes
Product Code. p-9393004
Click to view available options
Capacity (Metric):
2 mL
Closure Color:
Red with White Ring
Unit Size:
Pack of 1200
Product Code. 13068662
Brand: Greiner Bio-One 454096
VACUETTE™ Z Serum Blood Collection Tubes are used in determinations for clinical chemistry, immunology and serology. Available in Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator. The inner wall of the tube is coated with silica particles which activate clotting when the tubes are turned.
  • Tubes are supplied in packs of 50 pieces
  • For venus blood collection
  • Primarily used for drawing blood for clinical chemistry, immunology and serology testing
  • Specially coated with microscopic silica particles, which activate the coagulation process
  • Made out of virtually unbreakable PET plastic
  • Provides improved safety and hygiene during specimen collection procedures with the advantage of being as clear as glass
  • Available in: Serum Clot Activator, Serum Sep Clot Activator and Serum Fast Separator
  • CE marked
TRUSTED_SUSTAINABILITY
Specifications
Product Type Blood Collection Tube
Additive Clot Activator
Sterility Sterile
Closure Type Premium Safety Twist Cap
Capacity (Metric) 2 mL
For Use With (Equipment) VACUETTE™ Blood Collection Needles
Closure Color Red with White Ring
Tube Size 13 x 75 mm
Quantity Case of 24
For Use With (Application) Blood Collection
Length (Metric) 75 mm
Diameter (Metric) Outer 13 mm