Learn More
Serum Amyloid A1 Antibody (CL9175), Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-21169-100ul
 View more versions of this product
Product Code. 18612315
  • 5830.00 NOK / 100µL
Description
Serum Amyloid A1 Monoclonal antibody specifically detects Serum Amyloid A1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Serum Amyloid A1
Monoclonal
Unconjugated
PBS, pH 7.2, 40% glycerol
Mouse
Protein A purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
CL9175
Western Blot 1 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
MGC111216, PIG4, SAA, SAA2, serum amyloid A protein, serum amyloid A1, TP53I4, tumor protein p53 inducible protein 4
This antibody was developed against a Synthetic Peptide of P0DJI8. Actual immunogen sequence is proprietary.
100 μg
Alzheimers Research, Biologically Active Proteins, Cancer, Neuroscience
6288
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1