missing translation for 'onlineSavingsMsg'
Learn More

Sertaconazole nitrate, MedChemExpress

Product Code. p-200101589
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16458436

Brand: Medchem Express HYB0736A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sertaconazole nitrate (FI7056) is a broad-spectrum topical antifungal agent, exhibits anti-inflammatory activity via activation of a p38-COX-2-PGE2 pathway. Sertaconazole nitrate is also a microtubule inhibitor, shows antiproliferative effect, induces apoptosis and autophagy, and can also inhibit the migration of cells.

Sertaconazole nitrate (FI7056) is a broad-spectrum topical antifungal agent, exhibits anti-inflammatory activity via activation of a p38-COX-2-PGE2 pathway. Sertaconazole nitrate is also a microtubule inhibitor, shows antiproliferative effect, induces apoptosis and autophagy, and can also inhibit the migration of cells.

Specifications

Chemical Name or Material Sertaconazole nitrate
Physical Form Solid
CAS 99592-39-9
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C20H16Cl3N3O4S
For Use With (Application) COVID-19-immunoregulation
Synonym FI7056
Solubility Information DMSO : ≥ 100 mg/mL (199.69 mM)
SMILES ClC1=CC=C(C(OCC2=CSC3=C(Cl)C=CC=C32)CN4C=CN=C4)C(Cl)=C1.O[N+]([O-])=O
Molecular Weight (g/mol) 500.78
Formula Weight 500.78
Percent Purity 99.58%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.