missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SERPINH1 (Human) Recombinant Protein

Human SERPINH1 full-length ORF ( AAH14623.1, 1 a.a. - 418 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000871-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16142671

  • 4130.00 NOK / 10µg
Estimated Shipment: 06-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MRSLLLLSAFCLLEAALAAEVKKPAAAAAPGTAEKLSPKAATLAERSAGLAFSLYQAMAKDQAVENILVSPVVVASSLGLVSLGGKATTASQAKAVLSAEQLRDEEVHAGLGELLRSLSNSTARNVTWKLGSRLYGPSSVSFADDFVRSSKQHYNCEHSKINFRDKRSALQSINEWAAQTTDGKLPEVTKDVERTDGALLVNAMFFKPHWDEKFHHKMVDNRGFMVTRSYTVGVMMMHRTGLYNYYDDEKEKLQIVEMPLAHKLSSLIILMPHHVEPLERLEKLLTKEQLKIWMGKMQKKAVAISLPKGVVEVTHDLQKHLAGLGLTEAIDKNKADLSRMSGKKDLYLASVFHATAFELDTDGNPFDQDIYGREELRSPKLFYADHPFIFLVRDTQSGSLLFIGRLVRPKGDKMRDEL
Specifications

Specifications

AAH14623.1
serpin peptidase inhibitor, clade H (heat shock protein 47), member 1, (collagen binding protein 1)
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SERPINH1
GST
871
Wheat germ expression system
10 μg
AsTP3, CBP1, CBP2, HSP47, PIG14, PPROM, RA-A47, SERPINH2, gp46
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Videos
SDS
Documents
Certificates
Special Offers

Special Offers