Learn More
SERPING1/C1 Inactivator Mouse anti-Human, Unconjugated, Clone: 1B11B11, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66882-1-IG-150UL
 View more versions of this product
Product Code. 16899145
  • 4836.10 NOK / 150µL
Description
The SERPING1/C1 Inactivator antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SERPING1/C1 Inactivator. This antibody recognizes human, pig, rat antigen. The SERPING1/C1 Inactivator antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
SERPING1/C1 Inactivator
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Pig
Western Blot, ELISA
1B11B11
Unconjugated
150 μL
710
IgG2a