missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
The product encoded by this gene is a serine proteinase inhibitor which rapidly inhibits thrombin in the presence of dermatan sulfate or heparin. The gene contains five exons and four introns. This protein shares homology with antithrombin III and other members of the alpha 1-antitrypsin superfamily. Mutations in this gene are associated with heparin cofactor II deficiency.
Specifications
Specifications
| Antigen | SERPIND1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.13 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene | SERPIND1 |
| Gene Accession No. | P05546, P49182, Q64268 |
| Gene Alias | D22S673, HC II, HC2, HCF2, HCII, Heparin cofactor 2, Heparin cofactor II, HLS2, LS2, Protease inhibitor leuserpin 2, Serpin D1, SERPIND1 |
| Gene Symbols | Serpind1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?