missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ SERPINC1 (Human) Recombinant Protein

Product Code. 16121971
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
Product Code. Quantity unitSize
16121971 25 μg 25µg
16111971 10 μg 10µg
2 options
This item is not returnable. View return policy

Product Code. 16121971

Brand: Abnova™ H00000462P01.25ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human SERPINC1 full-length ORF ( AAH22309.1, 1 a.a. - 259 a.a.) recombinant protein with GST-tag at N-terminal.

  • Sequence: MYSNVIGTVTSGKRKVYLLSLLLIGFWDCVTCHGSPVDICTAEPRDIPMNPMCIYRSPEKKATEDEGSEQKIPEATNRRVWELSKANSRFATTFYQHLADSKNDNDNIFLSPLSISTAFAMTKLGACNDTLQQLMEVFKFDTISEKTSDQIHFFFAKLNCRLYRKANKSSKLVSANRLFGDKSLTFNDLYVSDAFHKAFLEVNEEGSEAAASTAVVIAGRSLNPNRVTFKANRPFLVFIREVPLNTIIFMGRVANPCVK

Specifications

Accession Number AAH22309.1
Gene ID (Entrez) 462
Name serpin peptidase inhibitor, clade C (antithrombin), member 1
Preparation Method Wheat germ expression system
Quality Control Testing 125% SDS-PAGE Stained with Coomassie Blue
Quantity 25 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias AT3, ATIII, MGC22579
Gene Symbol SERPINC1
Species Wheat Germ (in vitro)
Protein Tag GST
Buffer 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.