Learn More
Falcon™ Pipets, Serol Triple Bagged, Plastic/Plastic, Sterile, Bulk
Product Code. 30093042
Change view
Click to view available options
Volume (Metric):
1 mL
2 mL
5 mL
10 mL
25 mL
50 mL
Unit Size:
Pack of 1000
Pack of 200
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
30093042 25 mL Pack of 200
30093039 1 mL Pack of 1000
30093038 2 mL Pack of 1000
30093041 5 mL Pack of 200
30093040 10 mL Pack of 200
30093043 50 mL Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30093042 Supplier Falcon™ Supplier No. 355028
  • ONLINE EXCLUSIVE PRICE
    3120.00 NOK
    2848.46 NOK / Pack of 200
    Save 271.54 NOK
    8% Off
View all promotions
Estimated Shipment: 09-12-2026
Corning’s triple-bagged packaged pipets are a safety and convenience revolution. This packaging protects the product from external contamination and is ideal for laboratory clean rooms, such as cell-based vaccine or drug production facilities.
  • Triple-bagged in two outer bags
  • Manufactured and packaged with animal-free materials
  • Sterility Assurance Level (SAL) of 10-6
  • USP Class VI tested
  • Nonpyrogenic
  • Noncytotoxic
  • RNase-/DNase-free
  • Human DNA-free
  • Low particulate according to USP 788
  • Extractables data available upon request
  • Color-coded magnifier stripes make volume reading and size selection easier
  • Bidirectional graduations provide choice of ascending and descending scales
  • Negative graduations allow additional working volume
  • Each pipet is individually wrapped in easy open plastic
  • No need for decontamination spraying with alcohol
  • Manufactured in USA
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Sterility Sterile
Graduations Yes
Quantity 200/Pk.
Packaging Type Triple Bagged, Bulk
Color Transparent
Volume (Metric) 25 mL
Type Serological Pipet
Disposable Reusable
Unless otherwise specified, all products are for research use or general laboratory use only. Not intended for use in diagnostic or therapeutic procedures. Not for use in humans. These products are not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment. Corning Life Sciences makes no claims regarding the performance of these products for clinical or diagnostic applications.
Corning's products are not specifically designed and tested for diagnostic testing. Many Corning products, though not specific for diagnostic testing, can be used in the workflow and preparation of the test at the customers discretion. Customers may use these products to support their claims. We cannot make any claims or statements that our products are approved for diagnostic testing either directly or indirectly. The customer is responsible for any testing, validation, and/or regulatory submissions that may be required to support the safety and efficacy of their intended application.