Learn More
TPP Techno Plastic Products™ Serological Pipets
Product Code. 11361784
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
25 mL
50 mL
Graduations:
1/10
1/100
2/10
Color:
Blue
Green
Orange
Purple
Red
Unit Size:
Each
Pack of 50
Pack of 60
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Color unitSize
11361784 25 mL 2/10 Red Pack of 60
11321784 10 mL 1/10 Orange Each
11391774 2 mL 1/100 Green Each
11311784 5 mL 1/10 Blue Each
11341784 25 mL 2/10 Red Each
11371784 50 mL 1/10 Purple Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11361784 Supplier TPP Techno Plastic Products™ Supplier No. 94525
Manufactured with the highest grade polystyrene for robustness and extreme break resistance. TPP Techno Plastic Products™ Serological Pipets offers large capacity volumes in a compact, ergonomic design.
  • Accurate volume; highly visible, bidirectional (ascending, descending); over-volume graduations
  • Smooth interiors, prevent cell hang-up or clumping inside the pipette
  • Excellent optical transparency
  • Color coded cotton plugs and dispenser boxes for easy identification
  • Convenient, stackable TPP dispenser box
  • Single wrapped in lint-free paper/plastic blister
  • Slender and short form enables ergonomic working conditions in the sterile hood
  • Small outside diameter of the tube at the tip results in an excellent accessibility of the large pipettes even in small tissue culture flasks
  • Highly drop free pipet tip design
  • Optimized form of the mouthpiece preserves the rubber of the pipet aid
  • Models available with reservoir; reservoir augments the total volume capacity without adding proportionate length to the pipet
Specifications
Material Polystyrene
Graduated Yes
Graduations 2/10
Color Red
Volume (Metric) 25 mL