Learn More
Ohaus™ Series MB Reusable Pan
Shop All Ohaus Products
Click to view available options
:
Weight: 0.269 kg
Description
- Accessory
- For use with moisture analyzer
Specifications
Specifications
| Width (Metric) | 7 mm |
| Languages | Chinese, English (US), French, German, Italian, Japanese, Korean, Polish, Portuguese (BR), Russian, Spanish (MX) |
| Quantity | 3/Pk. |
| Weight (Metric) | 0.269 kg |