Learn More
Kautex™ HDPE, Series 353 Industrial Jerry Can
For Use With: Storage, packaging, transport; Color: Natural
Brand:  Kautex™ 2000080289
Product Code. 15921441
  • ONLINE EXCLUSIVE PRICE
    1195.00 NOK
    1162.74 NOK / Pack of 10
    Save 32.26 NOK
    2% Off
Only null left
Estimated Shipment: 03-02-2026
Description
  • Manufactured from HDPE
  • With UN approval on the 25 L model
  • Can be used for almost any purpose
  • Applications include storage, packaging and transport
  • Available in blue or natural color
  • Wide range of closure accessories available
  • Leakproof