Learn More
Robu™ Series 15 00 Porosity Filter Plates
Brand:  Robu™ 15409
Additional Details : Weight : 0.08170kg
 View more versions of this product
Product Code. 16822776
  • 695.00 NOK / Pack of 10
Estimated Shipment: 14-06-2024
Specifications
1Pack
Filter Plate
4.5 mm
40 mm