Learn More
Robu™ Series 15 00 Porosity Filter Plates
Brand:  Robu™ 15600
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12000442
  • 745.00 NOK / Pack of 5
Estimated Shipment: 24-06-2024
Specifications
1Pack
Filter Plate
5 mm
60 mm