Learn More
SERCA2 ATPase Antibody (IID8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 6 publications
Brand:  Novus Biologicals NB300-529-0.025ml
 View more versions of this product
Product Code. 18548890
  • 1965.00 NOK / 25µL
Description
SERCA2 ATPase Monoclonal specifically detects SERCA2 ATPase in Human, Mouse, Rat, Porcine, Amphibian, Bovine, Canine, Guinea Pig, Hamster, Primate, Rabbit, Sheep, Virus samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Block/Neutralize.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SERCA2 ATPase
Monoclonal
1 mg/mL
Western Blot 1:500 - 1:5000, Flow Cytometry 1 μg 106 cells, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:200, Immunohistochemistry-Paraffin 1 μg/mL, Immunohistochemistry-Frozen 1:500, Block/Neutralize
ATP2BFLJ20293, ATPase, Ca++ dependent, slow-twitch, cardiac muscle-2, ATPase, Ca++ transporting, cardiac muscle, slow twitch 2, Calcium pump 2, Calcium-transporting ATPase sarcoplasmic reticulum type, slow twitch skeletalmuscle isoform, cardiac Ca2+ ATPase, DAR, DD, EC 3.6.3, EC 3.6.3.8, Endoplasmic reticulum class 1/2 Ca(2+) ATPase, FLJ38063, MGC45367, sarcoplasmic/endoplasmic reticulum calcium ATPase 2, SERCA2DKFZp686P0211, SR Ca(2+)-ATPase 2
Mouse
Affinity Purified
RUO
488
Human
IgG1
Western Blot
IID8
Unconjugated
O46674
ATP2A2
Purified canine cardiac sarcoplasmic reticulum.
0.025 mL
Primary
Detects Sarcoplasmic or Endoplasmic Reticulum Calcium 2 (SERCA2) ATPase. This does not discriminate between the two isoforms.
Store at -20°C. Avoid freeze/thaw cycles.