Description
Biotin Binding Capacity of 2500 to 3500pmol/mg beads
Specifications
Specifications
| Concentration | 1% solids (10mg/mL) |
| Content And Storage | 2°C to 8°C |
| For Use With (Application) | Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets |
| Quantity | 1 mL |
| Diameter (Metric) | 1 μm |