Learn More
Cytiva Sera-Mag™ Streptavidin Magnetic Beads, high biotin binding capacity
Product Code. 11815022 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
Unit Size:
Each
Product Code. Quantity unitSize
11815022 100 mL Each
11805022 5 mL Each
11825022 1 mL Each
3 options
Product Code. 11815022
Brand: Cytiva 30152105010350
Their cauliflower-like structure gives Sera-Mag beads high binding capacity. Sera-Mag Streptavidin beads contain covalently bound streptavidin and are available with low, medium, or high nominal biotin binding capacities. Choose the biotin-binding capacity needed to optimize your application.
Biotin Binding Capacity of 4500 to 5500pmol/mg beads
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Immunoassays, cDNA Library Construction, Capture of Biotinylated Targets
Quantity 100 mL
Diameter (Metric) 1 μm