Learn More
Cytiva Sera-Mag SpeedBeads™ Protein A/G Magnetic Beads
Product Code. 15732442 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
Unit Size:
100mL
1mL
5mL
Product Code. Quantity unitSize
15732442 5 mL 5mL
15742442 100 mL 100mL
15752442 1 mL 1mL
3 options
Product Code. 15732442
Brand: Cytiva 17152104010150
Provide a fast, convenient method for manual and automated magnetic isolation of IgA and IgG proteins using affinity binding. The beads can be used with the broadest range of antibody species and subclasses (even when subclass identity has not been determined).
  • Their cauliflower-like structure gives Sera-Mag beads high binding capacity
  • Fast and easy IgA purification and IgG purification
TRUSTED_SUSTAINABILITY
Specifications
Concentration 1% solids (10mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) Affinity Purification
Quantity 5 mL
Diameter (Metric) 1 μm