Learn More
Cytiva Sera-Mag™ Carboxylate modified Magnetic Beads, hydrophobic
Product Code. 11889782 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
15 mL
100 mL
1 L
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11889782 1 L Each
13273589 15 mL Each
11879782 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11889782 Supplier Cytiva Supplier No. 44152105050450
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent for coupling oligos or proteins directly to beads. Their cauliflower-like structure gives Sera-Mag beads high binding capacity. The carboxyl groups on these single shell magnetite beads can be activated by standard EDC/EDAC chemistry to react with free amino groups, forming amide bonds.
  • High binding capacity
  • 1.0μm diameter
  • High carboxyl acid content for binding for binding DNA and proteins
  • Hydrophobic bead based on crosslinking requirements for the encapsulation layer
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5% solids (50mg/mL)
Content And Storage 2°C to 8°C
For Use With (Application) DNA Sample Prep, Protein Purification
Quantity 1 L
Diameter (Metric) 1 μm