Learn More
VELP Scientifica SER 158 Series Automatic Solvent Extractor
Product Code. p-200106362 Shop All Velp Products
Change view
Click to view available options
Dimensions (D x W x H):
358 x 546 x 570 mm
546 x 546 x 570 mm
Unit Size:
Each
Product Code. Dimensions (D x W x H) unitSize
17587566 546 x 546 x 570 mm Each
15783619 358 x 546 x 570 mm Each
2 options
Product Code. 17587566
Brand: VELP Scientifica S303A0380
This item requires a price update prior to being ordered.
Request a Price for this item here.
Fully automatic and cloud-enabled hot solvent extractor, for safety, fast, accurate, and reliable extractable matter determination.
FAST EXTRACTION
The automatic extraction process completes fully unattended analysis according to the Randall method that is five times faster than traditional Soxhlet. The high-speed heating plates in glass-ceramic ensure fast solvent boiling.
MINIMUM SOLVENT CONSUMPTION
The patent pending titanium condensers guarantee the unparalleled solvent recovery of more than 90% that is collected in cooled recovery tank.
HIGH PRODUCTIVITY AND VERSATILITY
The powerful ControlPad is able to control up to 4 SER 158 units (3 or 6 positions) independently allowing to increase the number of units according to throughput requirement.
MAXIMUM SAFETY
  • SolventXpress™ ensures zero exposure to solvents
  • SafeEnd™ prevents the overheating and burning of soluble matter
  • Remote analysis interruption and immediate notifications trough VELP Ermes
EASE OF USE
The SER 158 Series makes hot solvent extraction easy and fast:
  • Preset library of methods and possibility to add new ones and mark preferred methods
  • Multilingual support Balance connection for automatic result calculation
  • Barcorde reader to reduce risk of errors
SMART INSTRUMENT
The SER 158 is designed to work unattended 24/7 with Load and Go technologies, minimizing necessary operator intervention. Thanks to VELP Ermes connection is possible to monitor the analysis and be notified, anytime, anywhere.
Specifications
Type Accelerated Solvent Extractor
Dimensions (D x W x H) 546 x 546 x 570 mm
Voltage 115/230 V