Learn More
Fisherbrand™ Septum for 11mm Crimp Cap
Most septa are laminated on one side with PTFE for high chemical resistance and an inert barrier between sample and carrier material.
Brand:  Fisherbrand™
Additional Details : Weight : 0.18000kg
 View more versions of this product
Product Code. 10041204
  • 2290.00 NOK / Pack of 1000
Estimated Shipment: 10-05-2024
Description
  • 11mm Septa
Specifications
Specifications
Septa
11 mm
1000
Red, White
PTFE, Silicone
1.3 mm