Learn More
- Home
- Products
- Chromatography Supplies
- Chromatography Septa
- Thermo Scientific™ Septa
Thermo Scientific™ Septa
Product Code. 11998409 Shop All Thermo Scientific Products| Product Code. | Description | Diameter (Metric) Inner | Temperature (Metric) Max | Quantity | Unit Size |
|---|---|---|---|---|---|
| 11998409 | TR-Green | 17 mm | 300°C | 50 (Glass Jar) | Pack of 50 |
| 10641324 | BTO | 17 mm | 300°C | 50 (Glass Jar) | Pack of 50 |
| 10513035 | BTO | 12.7 mm | 400°C | 50 (Glass Jar) | Pack of 50 |
| 10293553 | BTO | 11.5 mm | 400°C | 50 (Glass Jar) | Pack of 50 |
| 11908419 | TR-Green | 12.7 mm | 350°C | 50 (Glass Jar) | Pack of 50 |
| 11918419 | TR-Green | 11 mm | 350°C | 50 (Glass Jar) | Pack of 50 |
| 11928419 | TR-Green | 9 mm | 350°C | 50 (Glass Jar) | Pack of 50 |
| 10384563 | BTO | 9 mm | 400°C | 50 (Glass Jar) | Pack of 50 |
| 15320431 | BTO | 11.5 mm | 400°C | 50 (Blister Pk.) | Pack of 50 |
| 15340431 | BTO | 11 mm | 400°C | 50 (Blister Pk.) | Pack of 50 |
| 15360431 | BTO | 9 mm | 400°C | 50 (Blister Pk.) | Pack of 50 |
| 15300431 | BTO | 12.7 mm | 400°C | 48 (Blister Pk.) | Pack of 48 |
| 15294543 | TR-Blue | 17 mm | 200°C to 250°C | 50 (Glass Jar) | Each |
| 15274543 | TR-Blue | 11.5 mm | 200°C to 250°C | 50 (Glass Jar) | Each |
| 15264543 | TR-Blue | 11 mm | 200°C to 250°C | 50 (Glass Jar) | Each |
| 15254543 | TR-Blue | 9 mm | 200°C to 250°C | 50 (Glass Jar) | Each |
| 15380431 | TR-Green | 17 mm | 300°C | 48 (Blister Pk.) | Pack of 48 |
| 15320441 | TR-Green | 11 mm | 350°C | 50 (Blister Pk.) | Pack of 50 |
| 15244543 | Marathon | 17 mm | 300°C | 50 (Glass Jar) | Each |
| 15360441 | Marathon | 17 mm | 300°C | 48 (Blister Pk.) | Pack of 48 |
| 15380441 | Marathon | 11.5 mm | 350°C | 50 (Blister Pk.) | Pack of 50 |
| 15300451 | Marathon | 11 mm | 350°C | 50 (Blister Pk.) | Pack of 50 |
| 15320451 | Marathon | 9 mm | 350°C | 50 (Blister Pk.) | Pack of 50 |
| 15340441 | TR-Green | 9 mm | 350°C | 50 (Blister Pk.) | Pack of 50 |
Use promo code 27992
Log in to see stock.
BEST PRICE on Promo! Use promo code "27992" to get your promotional price.
Description
Ensure optimal performance of your GC instrument with bleed and temperature optimized Thermo Scientific™ BTO and TR-Green Septa. Made of low-bleed silicone, these septa have excellent mechanical properties, are ideal for demanding GC-MS applications, and may be used reliably up to 400 °C.
- Variety of materials and temperature limits for all applications
- For use in Thermo Scientific™ TRACE™ and FOCUS™ GCs and instruments from other manufacturers
- Contaminant-free in individual blister packs
Specifications
Specifications
| Product Type | Low Coring Septum |
| Description | TR-Green |
| Temperature (Metric) Max | 300°C |
| For Use With (Equipment) | Trace and Focus GC Instruments |
| Material | Silicone |
| Diameter (Metric) Inner | 17 mm |
| Quantity | 50 (Glass Jar) |