Learn More
Macherey-Nagel™ Septa
Brand: Macherey-Nagel™ 70277
Additional Details : Weight : 0.12600kg
Specifications
Silver | |
Butyl red/ PTFE gra | |
100 Pk. | |
Crimp Cap |
Aluminum, Butyl Rubber, PTFE | |
9.1 mm | |
3 mm |