missing translation for 'onlineSavingsMsg'
Learn More

Origene Technologies SEPT9 Monoclonal Antibody (OTI3G2), TrueMAB™, OriGene

Mouse Monoclonal Antibody

Brand:  Origene Technologies TA812811

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 17557424

  • 5484.49 NOK / 100µL
Estimated Shipment: 19-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SEPT9 Monoclonal antibody specifically detects SEPT9 in Human samples. It is validated for Western Blot
Specifications

Specifications

SEPT9
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UHD8
Mouse
Protein A/G
RUO
10801
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
OTI3G2
Unconjugated
SEPTIN9
AF17q25; eighth septin; Eseptin; E-septin; KIAA0991; MLL septin-like fusion; MLL septin-like fusion protein; MLL septin-like fusion protein MSF-A; MSF; MSF1; NAPB; Ov/Br septin; ovarian/breast septin; PNUTL4; SEPT9; SeptD1; septin 9; septin D1; SEPTIN9; Septin-9; septin-like protein; Sint1; SL3-3 integration site 1 protein; SLP; Slpa
Full length human recombinant protein of human SEPT9 (NP_006631) produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers