missing translation for 'onlineSavingsMsg'
Learn More

SEPT9 Monoclonal Antibody (OTI3G2), TrueMAB™, OriGene

Product Code. 17577424
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
Unit Size:
100µL
30µL
Product Code. Quantity unitSize
17577424 30 μL 30µL
17557424 100 μL 100µL
2 options
This item is not returnable. View return policy

Product Code. 17577424

Brand: Origene Technologies TA812811S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SEPT9 Monoclonal antibody specifically detects SEPT9 in Human samples. It is validated for Western Blot

Specifications

Antigen SEPT9
Applications Western Blot
Classification Monoclonal
Clone OTI3G2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SEPTIN9
Gene Accession No. Q9UHD8
Gene Alias AF17q25; eighth septin; Eseptin; E-septin; KIAA0991; MLL septin-like fusion; MLL septin-like fusion protein; MLL septin-like fusion protein MSF-A; MSF; MSF1; NAPB; Ov/Br septin; ovarian/breast septin; PNUTL4; SEPT9; SeptD1; septin 9; septin D1; SEPTIN9; Septin-9; septin-like protein; Sint1; SL3-3 integration site 1 protein; SLP; Slpa
Host Species Mouse
Immunogen Full length human recombinant protein of human SEPT9 (NP_006631) produced in HEK293T cell.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10801
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.