missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEPT14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24590-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
SEPT14 is a member of the highly conserved septin family of GTP-binding cytoskeletal proteins implicated in membrane transport, apoptosis, cell polarity, cell cycle regulation, cytokinesis, and other cellular functions.[supplied by OMIM, Jul 2008].Specifications
SEPT14 | |
Polyclonal | |
Unconjugated | |
SEPT14 | |
SEPT14, septin 14 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
346288, 689118, 74222 | |
-20°C | |
Liquid |
Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q6ZU15, Q9DA97 | |
SEPT14 | |
SEPT14 Fusion Protein Ag20181 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |