Description
SEPT11 belongs to the conserved septin family of filament-forming cytoskeletal GTPases that are involved in a variety of cellular functions including cytokinesis and vesicle trafficking.
Specifications
Specifications
| Antigen | SEPT11 |
| Applications | Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.35 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SEPT11 |
| Gene Accession No. | B3GNI6, Q8C1B7, Q9NVA2 |
| Gene Alias | SEPT11, septin 11 |
| Gene Symbols | SEPT11 |
| Show More |