Learn More
asecos™ Separation Barrier
Product Code. 17846376 Shop All asecos Products
Click to view available options
Length (Metric):
2 m
3 m
400.00 mm
Unit Size:
Each
Product Code. 17846376
Brand: asecos™ 13093
This item requires a price update prior to being ordered.
Request a Price for this item here.
The Separation Barrior provides rapid protection in case of leaking liquids.
  • Stick to ground and protects the environment from escaping liquids (e.g. for drains).
  • Consists of a special plastic mixture.
  • Resistant to water, hydrocarbon and most chemicals.
  • Reusable (can be washed with soapy water or detoxicated).
  • For all smooth substrates.
Specifications
Color Yellow
Material Polyurethane
Type Barrier Spillblocker
Length (English) 6.56 ft.
Length (Metric) 2 m
Width (English) 1.77 in.
Width (Metric) 45.00 mm
Height (English) 1.97 in.
Height (Metric) 50.00 mm
Weight (English) 8.38 lb.
Weight (Metric) 3.8 kg