Learn More
Lenz Laborglasinstrumente™ Separating Funnels with PTFE Plug
Product Code. 10316121
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Each
Product Code. 10316121
Brand: Lenz Laborglasinstrumente™ 04030173
Only null left
Add to basket
Add to basket
Made from Borosilicate glass. Lenz Laborglasinstrumente™ Dropping Funnels with PTFE Plug are fitted with a stopcock and are excellent for transferring fluids.
  • NS Socket and PE stopper
  • Without scale, with PTFE plug
  • Inscription label
  • Borosilicate glass
Specifications
Capacity (Metric) 2000 mL
Material Borosilicate Glass
Pore Size 6.0 mm
Quantity 1
Stopper Material Polyethylene