Learn More
Lenz Laborglasintrumente Separating Funnels, Conical, Amber, without Scale, with PTFE Plug
Product Code. 11993441
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
500 mL
1 L
2 L
Unit Size:
Each
Pack of 2
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
11993441 500 mL Pack of 2
11903451 2 L Each
11913451 1 L Pack of 2
11963441 50 mL Pack of 2
11973441 100 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 11993441 Supplier Lenz Laborglasintrumente Supplier No. 04008158
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • With NS socket and PE stopper
  • Without scale, with PTFE plug
  • With screwthread retaining device
  • Material: borosilicate glass 3.3
Specifications
Capacity (Metric) 500 mL
Material Borosilicate Glass (3.3)