Learn More
Duran™ Separating Funnel Taps With Short funnel
Shop All DWK Life Sciences Products
Click to view available options
Description:
Stopper No.: NS 12.5
Stopper No.: NS 14.5
Stopper No.: NS 18.8
Unit Size:
Pack of 25
Pack of 50
Specifications
Specifications
| Type | Separating Funnel Tap |
| Description | Stopper No.: NS 14.5 |
| Material | Glass |
| Includes | Short funnel |