Learn More
Cytiva Sentino™ Microbiology Pump Replacement Tubing Set and Adapter
Description
- For vacuum filtration of liquids on our Sentino pump
- For the analysis of aqueous samples for microbiological and particulate contamination using the Membrane Filter (MF) Technique
- For research use only
Specifications
Specifications
| For Use With (Equipment) | MicroFunnel™ Filter Funnel |
| Sterility | Sterile |
| No. per Pack | 10/Pk. |