Learn More
Cytiva Sentino™ Metricel™ Black Membrane
Product Code. 17184591
Change view
Click to view available options
Pore Size:
0.45 μm
0.8 μm
Unit Size:
Pack of 200
Product Code. Pore Size unitSize
17184591 0.45 μm Pack of 200
17194591 0.8 μm Pack of 200
2 options
Product Code. 17184591
Brand: Cytiva 68124
Only null left
Add to basket
Add to basket
Filter dispense packs for Sentino dispenser are ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes.
  • Filter dispensers are ideal for high throughput microbiology testing labs looking for more ergonomic, faster ways to handle individual membranes
  • Designed to aseptically dispense one disc of membrane at the touch of a button
  • Gamma irradiated and ready to use
  • For research use only
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material (Membrane) Modified PES
Pore Size 0.45 μm
Sterility Non-sterile
No. per Pack 1000/Pk., Dispenser Pack of 200
Diameter (Metric)Top 47 mm