Learn More
Cytiva Sentino™ Global Membrane Dispenser
Product Code. 17184251
Change view
Click to view available options
:
Automatic
Product Code.
17184251 Automatic
1 options
Product Code. 17184251
Brand: Cytiva 13184
Only null left
Add to basket
Add to basket
Sentino Dispenser aseptically dispenses individual membrane filters with a single press on its soft-touch keypad.
  • Sentino range is a complete system of products for evaluation of microbial contamination of aqueous samples using the Membrane Filter (MF) Technique
TRUSTED_SUSTAINABILITY
Specifications
Sterility Non-sterile
Includes NiMH nickel metal hydride battery
Type Automatic Dispenser
Length (Metric) 22 cm
Width (Metric) 12 cm
Height (Metric) 20 cm