Learn More
Cytiva Sentino™ Funnel Housing
Product Code. 17154281
Change view
Click to view available options
:
300 mL
Product Code.
17154281 300 mL
1 options
Product Code. 17154281
Brand: Cytiva 4243
Only null left
Add to basket
Add to basket
Magnetic funnels replacement housing designed for use with our Sentino pump.
  • Unique magnetic seal allows easy, one-handed vacuum filtration of liquids with compatible membranes
  • Not approved for use in medical, clinical, surgical or other patient applications
TRUSTED_SUSTAINABILITY
Specifications
Material Polyphenylsulfone
Sterility Non-sterile
Volume (Metric) 300 mL