Få mere at vide
Thermo Scientific™ Sensititre™ EU Surveillance ESBL EUVSEC2 AST Plate
Beskrivelse
Comply with EU Surveillance System program for ESBL isolates with this convenient plate.
Perform accurate AST with ESBL isolates specific formulary as a part of EU Surveillance program with Thermo Scientific™ Sensititre EU Surveillance ESBL EUVSEC2 AST Plate.
Comply with EU Surveillance System program for ESBL isolates with Sensititre EU Surveillance EU Surveillance ESBL EUVSEC2 AST Plate.
- Extended shelf life, room temperature storage eliminates inventory control concerns
- Individual packaging allows a laboratory to test one plate at a time with no waste
- Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
- Automated or manual read options are available for ultimate flexibility
Antimicrobial Resistance (AMR) is a significant concern and of increasing importance. The inappropriate use of antimicrobial agents in humans and animals is leading this growing and dangerous issue, emerging as a public health threat. This is why the European Parliament, the Council and its Agencies: European Centre for Disease Prevention and Control (ECDC), the European Food Safety Authority (EFSA) and the European Medicines Agency (EMA), have identified the need to establish a common strategy in the fight against AMR.
The Sensititre System utilizes true minimum inhibitory concentration (MIC) results, which are essential to identifying antimicrobial resistance, and provide greater sensitivity for better resistance tracking.
With a wide range of Sensititre plates specifically designed for surveillance programs, the Sensititre System is a leader in antimicrobial susceptibility testing for surveillance.
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand
Tekniske data
Tekniske data
| Description | Sensititre EU Surveillance ESBL EUVSEC2 AST Plate |
| Product Type | AST Plate |
| Test Type | Antimicrobial Surveillance, Minimum Inhibitory Concentration (MIC) |
| Isolates Per Plate | 1 |
| No. per Pack | 10 |
| Packaging Type | Box |
| Quantity | 10 Plates/Box |
| Reading Options | Manual, Vizion |
| Regulatory Status | RUO |
| Specimen Type | ESBL Isolates |
| Vis mere |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.