Learn More
Ansell™ HyFlex™ 48-105 Series White Polyurethane Light Weight Gloves
Product Code. 12690922 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12690922 11 12 pairs
12680922 10 12 pairs
12600932 6 12 pairs
12630932 9 12 pairs
12610932 7 12 pairs
12620932 8 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12690922 Supplier Ansell™ Supplier No. 48105110
  • ONLINE EXCLUSIVE PRICE
    492.00 NOK
    323.86 NOK / 12 pairs
    Save 168.14 NOK
    34% Off
 Restricted Item
Estimated Shipment: 15-12-2026
Designed for light applications that require good levels of comfort and abrasion resistance.
  • Industrial gloves
  • Comfort and protection for lightweight applications
  • Offers precise handling of small objects in dry or light oily environments
  • Provides enhanced safety, dexterity and durability
Applications:
  • Assembling, inspecting and packing small components
  • Light assembly tasks
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance Standard CATEGORY II EN 388 : 0030
Cuff Material Nylon
Accelerator Free Yes
Applications Industrial
Color White
Disposable Reusable
Exterior Finish No Texture
Latex Free Yes
Material Polyurethane
Powdered Powder Free
Product Type Lightweight Gloves
Size 11
Sterility Non-sterile
Glove Cuff Color White
Glove Cuff Style Knit
Liner Material Nylon
Lining Lined
Quantity 12 Pairs
Resistant To Abrasion resistance
Series 48-105