Learn More
Ansell™ HyFlex™ 48-105 Series White Polyurethane Light Weight Gloves
Product Code. 12630932 Shop All Ansell Products
Change view
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. Size unitSize
12630932 9 12 pairs
12680922 10 12 pairs
12600932 6 12 pairs
12690922 11 12 pairs
12610932 7 12 pairs
12620932 8 12 pairs
6 options
Product Code. 12630932
Brand: Ansell™ 48105090
Only null left
Add to basket
Add to basket
Designed for light applications that require good levels of comfort and abrasion resistance.
  • Industrial gloves
  • Comfort and protection for lightweight applications
  • Offers precise handling of small objects in dry or light oily environments
  • Provides enhanced safety, dexterity and durability
Applications:
  • Assembling, inspecting and packing small components
  • Light assembly tasks
TRUSTED_SUSTAINABILITY
Specifications
Back Material Polyurethane
Certifications/Compliance Standard CATEGORY II EN 388 : 0030
Cuff Material Nylon
Accelerator Free Yes
Applications Industrial
Color White
Disposable Reusable
Exterior Finish No Texture
Latex Free Yes
Material Polyurethane
Powdered Powder Free
Product Type Lightweight Gloves
Size 9
Sterility Non-sterile
Glove Cuff Color White
Glove Cuff Style Knit
Liner Material Nylon
Lining Lined
Quantity 12 Pairs
Resistant To Abrasion resistance
Series 48-105