Learn More
Semperit™ Sempermed™ Supreme Natural Rubber Latex Surgical Gloves
Product Code. p-8481019
Click to view available options
Size:
8
Unit Size:
50 pairs
Product Code. 10752391
Brand: Semperit™ 3000006094
Only null left
Add to basket
Add to basket
Powder-free surgical gloves ideal for double-donning. Provides optimum safety and ideal tactile properties.
  • Patented synthetic lining makes these gloves considerably easy to don.
  • Provides optimal tactile sensitivity.
  • Developed specifically for the requirements of the operating theater, the fully anatomical shape with curved fingers corresponds with the natural, relaxed hand posture.
Specifications
Certifications/Compliance EN 1041, EN 980, EN 455/3, ISO 15223
Color White
Disposable Single-use
Material Latex
Powdered Powder Free
Size 8
Sterility Sterile
Thickness (Metric) 0.19 to 0.24 mm
Glove Cuff Style Rolled
Length (Metric) 280 mm