Learn More
Julabo™ SemiChill Recirculating Cooling Baths with Air Compressor
Product Code. 11914838
Change view
Click to view available options
Capacity (Metric):
21 to 33 L
43 to 60 L
Unit Size:
Each
Product Code. Capacity (Metric) unitSize
11914838 21 to 33 L Each
11934838 43 to 60 L Each
2 options
Product Code. 11914838
Brand: Julabo™ 950002503P3H0
Only null left
Add to basket
Add to basket
Operational temperature ranges from +5 to +35 °C. Julabo™ SemiChill Recirculating Cooling Baths with Air Compressor can be modified to provide heater capacities up to 5 kW.
  • Space-saving, no lateral ventilation
  • Handles and castor make relocation easy
  • Industrial grade mains power switch and emergency cut-off
  • Pressure gauge
  • Liquid level indicator
  • Front filling port
  • Low noise level
  • PID temperature control
  • ATC3 3-point calibration
  • Early warning system for low liquid level
  • Warning and safety functions
  • Modular design allows selection between different options
  • Air cooled model
Specifications
Capacity (Metric) 21 to 33 L
Certifications/Compliance CE, Classification III (FL) according to DIN 12876-1
Cooling Capacity 0.9kW at -10°C, 1.5kW at 0°C, 2.5kW at 20°C (Medium Ethanol)
Flow Rate 33 L/min.
For Use With (Equipment) For industrial applications
Temperature Range 5°C to 40°C
Heating Capacity 1 kW
Max. Pressure 50.8 psi
Model SC2500a
Refrigerant R404A
Temperature Stability ±0.1°C
Length (Metric) 62 cm
Width (Metric) 49 cm
Height (Metric) 105 cm
Weight (Metric) 123 kg
Electrical Requirements 230 V 60 Hz 10 A
Voltage 230 V
Frequency 60 Hz