Learn More
Semicarbazide Hydrochloride, Sigma-Aldrich™
Specifications
Specifications
| CAS | 563-41-7 |
| Melting Point | 175°C to 177°C (Decomposition) |
| MDL Number | MFCD00013009 |
| Quantity | 100 g |
| Synonym | Semicarbazide hydrochloride, Semicarbazide hydrochloride |
| Molecular Weight (g/mol) | 75.07 (Free Base Basis) |
| Grade | Research |