missing translation for 'onlineSavingsMsg'
Learn More

Proteintech Semenogelin-1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15916-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895314

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-06-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is the predomit protein in semen. The encoded secreted protein is involved in the formation of a gel matrix that encases ejaculated spermatozoa. The prostate-specific antigen (PSA) protease processes this protein into smaller peptides, with each possibly having a separate function. The proteolysis process breaks down the gel matrix and allows the spermatozoa to move more freely.
Specifications

Specifications

Semenogelin-1
Polyclonal
Unconjugated
SEMG1
Semenogelin 1, semenogelin I, Semenogelin-1, SEMG, SEMG1, SGI
Rabbit
Antigen Affinity Chromatography
RUO
6406
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04279
SEMG1
Semenogelin-1 Fusion Protein Ag8717
20 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers