Learn More
Semaphorin 4C Sheep anti-Human, Polyclonal, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody has been used in 3 publications
Brand:  R&D Systems AF6125-SP
Encompass
Product Code. 15778992
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Semaphorin 4C Polyclonal specifically detects Semaphorin 4C in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.