missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SEMA7A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18070-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
CD108 (Sema7A) is a GPI-anchored semaphorin family member, which enhances central and peripheral axonal growth and is required for proper axon track formation during embryogenesis. CD108 also regulates osteoclast differentiation and pre-osteoblastic cell migration, and in immune system affects cell proliferation, chemotaxis and cytokine release. On erythrocytes CD108 defines the JMH (John-Milton-Hagen) human blood group. CD108 signalizes through its receptors - plexin C1 and beta1 integrins.Specifications
SEMA7A | |
Polyclonal | |
Unconjugated | |
SEMA7A | |
CD108, CDw108, H SEMA K1, H Sema L, JMH, JMH blood group antigen, Sema K1, Sema L, SEMA7A, SEMAK1, SEMAL, Semaphorin 7A, Semaphorin K1, Semaphorin L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20361, 8482 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O75326, Q9QUR8 | |
SEMA7A | |
SEMA7A Fusion Protein Ag12707 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |