Learn More
SEMA4A Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27359-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16895585
  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.
Description
Description
The SEMA4A antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human SEMA4A. This antibody recognizes human antigen. The SEMA4A antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SEMA4A
Polyclonal
Unconjugated
150 μL
64218
IgG
Western Blot, Immunohistochemistry, ELISA
333 μg/mL