Learn More
Steris™ Self-Sealing sterilization Pouches
Product Code. 15448782
Change view
Click to view available options
Dimensions (L x W):
90 x 270mm
200 x 350mm
Unit Size:
Pack of 200
Product Code. Dimensions (L x W) unitSize
15448782 200 x 350mm Pack of 200
15428782 90 x 270mm Pack of 200
2 options
Product Code. 15448782
Brand: Steris™ NWKP2035
Only null left
Add to basket
Add to basket
Provides an effective means of sterilization, safe handling and storage of all items until the moment they are used. Steris Corporation™ Self-Sealing sterilization Pouches are CE marked for steam and EO gas.
  • Self-sealing bags eliminate the need for sealing devices
  • Excellent adhesive strength resistant to steam exposure
  • Non-toxic water-based indicator inks
  • Perforated flap for easy closing
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance CE Marked
For Use With (Equipment) Steam and Gas EO Sterilizers
Quantity 200/Pk.
Dimensions (L x W) 200 x 350mm
Length (Metric) 200 mm
Width (Metric) 350 mm