Learn More
- Home
- Products
- Sterilizers and Autoclaves
- Autoclave Bags, Wraps, and Sleeves
- Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 24 to 62 in. Wide
Keystone Cleanroom Products Self-Sealing 1421B Tyvek™ Autoclave Bag with Steam Indicator, 24 to 62 in. Wide
Product Code. 30020382
Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
12 x 34 in.
16 x 30 in.
16 x 44 in.
20 x 30 in.
24 x 24 in.
24 x 36 in.
24 x 40 in.
28 x 26 in.
28 x 28 in.
28 x 30 in.
28 x 36 in.
30 x 24 in.
30 x 30 in.
32 x 28 in.
32 x 32 in.
34 x 26 in.
34 x 28 in.
34 x 34 in.
36 x 24 in.
36 x 30 in.
36 x 36 in.
38 x 38 in.
40 x 24 in.
40 x 40 in.
42 x 36 in.
42 x 42 in.
44 x 28 in.
44 x 42 in.
46 x 26 in.
46 x 36 in.
48 x 24 in.
48 x 32 in.
48 x 36 in.
48 x 40 in.
48 x 48 in.
50 x 44 in.
50 x 48 in.
50 x 62 in.
52 x 24 in.
54 x 50 in.
56 x 34 in.
60 x 30 in.
60 x 36 in.
60 x 40 in.
62 x 24 in.
64 x 36 in.
64 x 48 in.
72 x 36 in.
72 x 40 in.
Unit Size:
Pack of 100
Pack of 125
Pack of 250
49 product options available for selection
| Product Code. | Dimensions (L x W) | Unit Size |
|---|---|---|
| 30020382 | 32 x 32 in. | Pack of 250 |
| 30020015 | 52 x 24 in. | Pack of 250 |
| 30020018 | 34 x 34 in. | Pack of 250 |
| 30020029 | 32 x 28 in. | Pack of 250 |
| 30020037 | 12 x 34 in. | Pack of 250 |
| 30020048 | 54 x 50 in. | Pack of 100 |
| 30020075 | 16 x 44 in. | Pack of 250 |
| 30020095 | 62 x 24 in. | Pack of 250 |
| 30020096 | 44 x 28 in. | Pack of 250 |
| 30020097 | 48 x 48 in. | Pack of 125 |
| 30020101 | 16 x 30 in. | Pack of 250 |
| 30020104 | 24 x 36 in. | Pack of 250 |
| 30020108 | 34 x 26 in. | Pack of 250 |
| 30020125 | 72 x 40 in. | Pack of 125 |
| 30020132 | 40 x 40 in. | Pack of 125 |
| 30020136 | 60 x 30 in. | Pack of 100 |
| 30020140 | 64 x 48 in. | Pack of 100 |
| 30020143 | 40 x 24 in. | Pack of 250 |
| 30020145 | 72 x 36 in. | Pack of 125 |
| 30020148 | 20 x 30 in. | Pack of 250 |
| 30020170 | 28 x 36 in. | Pack of 250 |
| 30020190 | 50 x 44 in. | Pack of 125 |
| 30020197 | 30 x 30 in. | Pack of 250 |
| 30020205 | 36 x 24 in. | Pack of 250 |
| 30020208 | 60 x 40 in. | Pack of 125 |
| 30020212 | 36 x 30 in. | Pack of 250 |
| 30020226 | 64 x 36 in. | Pack of 125 |
| 30020240 | 44 x 42 in. | Pack of 125 |
| 30020248 | 50 x 48 in. | Pack of 125 |
| 30020255 | 36 x 36 in. | Pack of 250 |
| 30020270 | 28 x 28 in. | Pack of 250 |
| 30020275 | 42 x 36 in. | Pack of 125 |
| 30020278 | 46 x 26 in. | Pack of 250 |
| 30020284 | 48 x 36 in. | Pack of 125 |
| 30020292 | 46 x 36 in. | Pack of 125 |
| 30020298 | 60 x 36 in. | Pack of 125 |
| 30020299 | 24 x 24 in. | Pack of 250 |
| 30020301 | 34 x 28 in. | Pack of 250 |
| 30020302 | 28 x 30 in. | Pack of 250 |
| 30020312 | 28 x 26 in. | Pack of 250 |
| 30020339 | 38 x 38 in. | Pack of 125 |
| 30020341 | 48 x 32 in. | Pack of 125 |
| 30020342 | 48 x 40 in. | Pack of 125 |
| 30020343 | 24 x 40 in. | Pack of 125 |
| 30020350 | 56 x 34 in. | Pack of 125 |
| 30020363 | 42 x 42 in. | Pack of 125 |
| 30020364 | 50 x 62 in. | Pack of 125 |
| 30020367 | 30 x 24 in. | Pack of 250 |
| 30020379 | 48 x 24 in. | Pack of 250 |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 49 options available.
This item is not returnable.
View return policy
This item is not returnable.
View return policy
Description
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Integrated steam indicator
- Recommended for sterilization of pharmaceutical equipment
- Manufactured and packaged in controlled cleanroom environment
- Eliminates need to cut material and the use of autoclave tape
- Self-sealing closure is fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Resists tearing and punctures
- Superior microbial barrier
- Certificates of conformance sent with shipment
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance (COC) |
| Closure Type | Self-seal |
| Color | White |
| Disposable | Single-use |
| For Use With (Application) | Steam Sterilization |
| For Use With (Equipment) | Pharmaceutical Equipment and Components |
| Includes | Steam Sterilization Indicator |
| Material | Tyvek |
| Sterility | Non-sterile |
| Temperature Range (English) | Up to 250°F |
| Temperature Range (Metric) | Up to 125°C |
| Thickness (English) | 5.3 mil |
| Dimensions (L x W) | 32 x 32 in. |
| Length (English) | 32 in. |
| Width (English) | 32 in. |
| Product Type | Autoclave Bag |
| Capacity (English) | 7.75 gal. |
| Capacity (Metric) | 29.3 L |
| Length (Metric) | 81 cm |
| Width (Metric) | 81 cm |
| Thickness (Metric) | 0.13 mm |
| Show More Show Less |