Learn More
Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
GREENER_CHOICE
Product Code. 17811711 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
3.5 x 8 in.
35.25 x 16 in.
6 x 2 in.
6 x 3 in.
8 x 3 in.
8 x 3.5 in.
8 x 6.125 in.
9 x 2.5 in.
10 x 5 in.
10 x 8 in.
12 x 6 in.
13 x 7 in.
15 x 4 in.
15 x 8.125 in.
18 x 8 in.
22 x 4 in.
28 x 6 in.
32 x 6 in.
34 x 8 in.
35 x 6 in.
4 x 15 in.
5 x 10 in.
6 x 35 in.
8 x 10 in.
8 x 15 in.
8 x 34 in.
10 x 11 in.
10 x 20 in.
12 x 10 in.
14 x 12 in.
15 x 20 in.
16 x 26 in.
16 x 32 in.
17 x 12 in.
17 x 16 in.
20 x 10.125 in.
20 x 15 in.
22 x 18 in.
24 x 14 in.
26 x 16 in.
26 x 24 in.
30 x 24 in.
32 x 16 in.
35 x 24 in.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. Dimensions (L x W) unitSize
17811711 10 x 20 in. Pack of 500
17841701 3.5 x 8 in. Pack of 500
17851701 4 x 15 in. Pack of 500
17861701 5 x 10 in. Pack of 500
17707827 6 x 2 in. Pack of 1000
17727827 6 x 3 in. Pack of 1000
17861711 6 x 35 in. Pack of 250
17737827 8 x 3 in. Pack of 500
17747827 8 x 3.5 in. Pack of 500
17787827 8 x 6.125 in. Pack of 500
17871701 8 x 10 in. Pack of 500
17881701 8 x 15 in. Pack of 500
17891701 8 x 34 in. Pack of 500
17717827 9 x 2.5 in. Pack of 1000
17777827 10 x 5 in. Pack of 500
17747837 10 x 8 in. Pack of 500
17801711 10 x 11 in. Pack of 500
17797827 12 x 6 in. Pack of 500
17787837 12 x 10 in. Pack of 500
17737837 13 x 7 in. Pack of 500
17707847 14 x 12 in. Pack of 500
17757827 15 x 4 in. Pack of 500
17757837 15 x 8.125 in. Pack of 500
17831711 15 x 20 in. Pack of 500
17841711 16 x 26 in. Pack of 500
17851711 16 x 32 in. Pack of 250
17717847 17 x 12 in. Pack of 500
17747847 17 x 16 in. Pack of 500
17767837 18 x 8 in. Pack of 500
17797837 20 x 10.125 in. Pack of 500
17737847 20 x 15 in. Pack of 500
17767827 22 x 4 in. Pack of 500
17750368 22 x 18 in. Pack of 500
17727847 24 x 14 in. Pack of 500
17757847 26 x 16 in. Pack of 500
17777847 26 x 24 in. Pack of 250
17707837 28 x 6 in. Pack of 500
17760368 30 x 24 in. Pack of 250
17717837 32 x 6 in. Pack of 500
17740368 32 x 16 in. Pack of 250
17777837 34 x 8 in. Pack of 500
17727837 35 x 6 in. Pack of 500
17787847 35 x 24 in. Pack of 250
17767847 35.25 x 16 in. Pack of 250
44 options
Product Code. 17811711
Brand: Keystone Cleanroom Products 01020TWBBSSSDSP02
Only null left
Add to basket
Add to basket
Self Seal Tyvek™ Steam Sterilization Pouches provide an excellent barrier from microbial contamination.
  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing an excellent barrier from microbial contamination
  • The clear film side offers excellent clarity along with strength and durability
  • These bags feature a medical grade adhesive strip for convenient self-sealing
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates the need for the use and validation of a Heat Sealer
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Self-seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Includes Steam Indicator Dot
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Quantity 25/Bag, 500/Cs.
Dimensions (L x W) 10 x 20 in.
Length (English) 10 in.
Length (Metric) 25.4 cm
Width (English) 20 in.
Width (Metric) 50.8 cm
Product Type Autoclave Bag