Learn More
Keystone Cleanroom Products Self Seal Tyvek™ Steam Sterilization Pouches
BIOPHARMAGREENER_CHOICE
Product Code. 17727847 Shop All Keystone Cleanroom Products Products
Change view
Click to view available options
Dimensions (L x W):
3.5 x 8 in.
35.25 x 16 in.
6 x 2 in.
6 x 3 in.
8 x 3 in.
8 x 3.5 in.
8 x 6.125 in.
9 x 2.5 in.
10 x 5 in.
10 x 8 in.
12 x 6 in.
13 x 7 in.
15 x 4 in.
15 x 8.125 in.
18 x 8 in.
22 x 4 in.
28 x 6 in.
32 x 6 in.
34 x 8 in.
35 x 6 in.
4 x 15 in.
5 x 10 in.
6 x 35 in.
8 x 10 in.
8 x 15 in.
8 x 34 in.
10 x 11 in.
10 x 20 in.
12 x 10 in.
14 x 12 in.
15 x 20 in.
16 x 26 in.
16 x 32 in.
17 x 12 in.
17 x 16 in.
20 x 10.125 in.
20 x 15 in.
22 x 18 in.
24 x 14 in.
26 x 16 in.
26 x 24 in.
30 x 24 in.
32 x 16 in.
35 x 24 in.
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. Dimensions (L x W) unitSize
17727847 24 x 14 in. Pack of 500
17841701 3.5 x 8 in. Pack of 500
17851701 4 x 15 in. Pack of 500
17861701 5 x 10 in. Pack of 500
17707827 6 x 2 in. Pack of 1000
17727827 6 x 3 in. Pack of 1000
17861711 6 x 35 in. Pack of 250
17737827 8 x 3 in. Pack of 500
17747827 8 x 3.5 in. Pack of 500
17787827 8 x 6.125 in. Pack of 500
17871701 8 x 10 in. Pack of 500
17881701 8 x 15 in. Pack of 500
17891701 8 x 34 in. Pack of 500
17717827 9 x 2.5 in. Pack of 1000
17777827 10 x 5 in. Pack of 500
17747837 10 x 8 in. Pack of 500
17801711 10 x 11 in. Pack of 500
17811711 10 x 20 in. Pack of 500
17797827 12 x 6 in. Pack of 500
17787837 12 x 10 in. Pack of 500
17737837 13 x 7 in. Pack of 500
17707847 14 x 12 in. Pack of 500
17757827 15 x 4 in. Pack of 500
17757837 15 x 8.125 in. Pack of 500
17831711 15 x 20 in. Pack of 500
17841711 16 x 26 in. Pack of 500
17851711 16 x 32 in. Pack of 250
17717847 17 x 12 in. Pack of 500
17747847 17 x 16 in. Pack of 500
17767837 18 x 8 in. Pack of 500
17797837 20 x 10.125 in. Pack of 500
17737847 20 x 15 in. Pack of 500
17767827 22 x 4 in. Pack of 500
17750368 22 x 18 in. Pack of 500
17757847 26 x 16 in. Pack of 500
17777847 26 x 24 in. Pack of 250
17707837 28 x 6 in. Pack of 500
17760368 30 x 24 in. Pack of 250
17717837 32 x 6 in. Pack of 500
17740368 32 x 16 in. Pack of 250
17777837 34 x 8 in. Pack of 500
17727837 35 x 6 in. Pack of 500
17787847 35 x 24 in. Pack of 250
17767847 35.25 x 16 in. Pack of 250
44 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 17727847
Marca: Keystone Cleanroom Products BBMGTVKSS1424
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Este artículo no se puede devolver. Vea la política de devoluciones
Self Seal Tyvek™ Steam Sterilization Pouches provide an excellent barrier from microbial contamination.
  • Great choice for equipment parts that are heavy, have sharp edges, or packaging that requires surface sanitization post-sterilization
  • DuPont Tyvek™/Polyester-based flexible film is lint-free, moisture and puncture resistant, extremely tough, and designed to be autoclaved
  • The Tyvek™ side offers high breathability while providing an excellent barrier from microbial contamination
  • The clear film side offers excellent clarity along with strength and durability
  • These bags feature a medical grade adhesive strip for convenient self-sealing
  • Each pouch has a CLASS A process indicator, which changes from pink to dark brown when exposed to steam
  • When the peel-able seals are opened, virtually no airborne particulates are generated
  • Recommended for sterilization of pharmaceutical equipment
  • Manufactured and packaged in controlled cleanroom environment
  • Eliminates the need for the use and validation of a Heat Sealer
Especificaciones
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance (COC)
Closure Type Self-seal
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Quantity 500/Pk.
Dimensions (L x W) 24 x 14 in.
Length (English) 24 in.
Length (Metric) 61 cm
Width (English) 14 in.
Width (Metric) 35.6 cm
Product Type Autoclave Bag
Mostrar más Mostrar menos
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.